Anti-FES Mouse Monoclonal Antibody [clone: 678527]

Supplier: R & D Systems
MAB6460
102886-432EA 577.02 USD
102886-432
Anti-FES Mouse Monoclonal Antibody [clone: 678527]
Antibodies
Type: Primary
Antigen Symbol: FES
Antigen Name: FES proto-oncogene, tyrosine kinase
Antigen Synonyms: FPS
Clonality: Monoclonal
Clone: 678527
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR