Anti-SYK Mouse Monoclonal Antibody [clone: 616268]

Supplier: R & D Systems
MAB6459
102886-430EA 633.42 USD
102886-430
Anti-SYK Mouse Monoclonal Antibody [clone: 616268]
Antibodies
Type: Primary
Antigen: SYK
Clonality: Monoclonal
Clone: 616268
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR