Anti-CRTC2 Rat Monoclonal Antibody [clone: 632307]

Supplier: R & D Systems
MAB6458
102886-428EA 561.11 USD
102886-428
Anti-CRTC2 Rat Monoclonal Antibody [clone: 632307]
Antibodies
Type: Primary
Antigen: CRTC2
Clonality: Monoclonal
Clone: 632307
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR