Anti-EBI3 Mouse Monoclonal Antibody [clone: 607201]

Supplier: R & D Systems
MAB6456
102886-426EA 600.15 USD
102886-426
Anti-EBI3 Mouse Monoclonal Antibody [clone: 607201]
Antibodies
Type: Primary
Antigen: EBI3
Clonality: Monoclonal
Clone: 607201
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR