Anti-GSTP1 Mouse Monoclonal Antibody [clone: 800027]

Supplier: R & D Systems
MAB6455
102886-424EA 594.37 USD
102886-424
Anti-GSTP1 Mouse Monoclonal Antibody [clone: 800027]
Antibodies
Simple Western

Type: Primary
Antigen Symbol: GSTP1
Antigen Name: glutathione S-transferase pi 1
Antigen Synonyms: DFN7, FAEES3, GST3, GSTP, HEL-S-22, PI
Clonality: Monoclonal
Clone: 800027
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR