Anti-WNT5A Rat Monoclonal Antibody [clone: 442625]

Supplier: R & D Systems
MAB645
220021-389EA 655.11 USD
220021-389
Anti-WNT5A Rat Monoclonal Antibody [clone: 442625]
Antibodies
Type: Primary
Antigen: WNT5A
Clonality: Monoclonal
Clone: 442625
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR