Anti-PRELP Mouse Monoclonal Antibody [clone: 754636]

Supplier: R & D Systems
MAB64471
102886-422EA 577.02 USD
102886-422
Anti-PRELP Mouse Monoclonal Antibody [clone: 754636]
Antibodies
Type: Primary
Antigen: PRELP
Clonality: Monoclonal
Clone: 754636
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR