Anti-PRELP Mouse Monoclonal Antibody [clone: 754633]

Supplier: R & D Systems
MAB6447
102886-420EA 584.25 USD
102886-420
Anti-PRELP Mouse Monoclonal Antibody [clone: 754633]
Antibodies
Type: Primary
Antigen: PRELP
Clonality: Monoclonal
Clone: 754633
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR