Anti-POGLUT1 Mouse Monoclonal Antibody [clone: 737512]

Supplier: R & D Systems
MAB64371
102886-208EA 561.11 USD
102886-208
Anti-POGLUT1 Mouse Monoclonal Antibody [clone: 737512]
Antibodies
Type: Primary
Antigen: POGLUT1
Clonality: Monoclonal
Clone: 737512
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR