Anti-POGLUT1 Mouse Monoclonal Antibody [clone: 737509]

Supplier: R & D Systems
MAB6437
102886-390EA 561.11 USD
102886-390
Anti-POGLUT1 Mouse Monoclonal Antibody [clone: 737509]
Antibodies
Type: Primary
Antigen: POGLUT1
Clonality: Monoclonal
Clone: 737509
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR