Anti-DGKI Mouse Monoclonal Antibody [clone: 648538]

Supplier: R & D Systems
MAB6435
102886-386EA 594.37 USD
102886-386
Anti-DGKI Mouse Monoclonal Antibody [clone: 648538]
Antibodies
Type: Primary
Antigen: DGKI
Clonality: Monoclonal
Clone: 648538
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR