Anti-VCAM1 Rat Monoclonal Antibody [clone: 112702]

Supplier: R & D Systems
MAB6434
220021-388EA 763.57 USD
220021-388
Anti-VCAM1 Rat Monoclonal Antibody [clone: 112702]
Antibodies
Type: Primary
Antigen: VCAM1
Clonality: Monoclonal
Clone: 112702
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR