Anti-AEBP1 Mouse Monoclonal Antibody [clone: 59831]

Supplier: R & D Systems
MAB6425
102886-384EA 561.11 USD
102886-384
Anti-AEBP1 Mouse Monoclonal Antibody [clone: 59831]
Antibodies
Type: Primary
Antigen: AEBP1
Clonality: Monoclonal
Clone: 59831
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR