Anti-NUDT5 Mouse Monoclonal Antibody [clone: 739044]

Supplier: R & D Systems
MAB6414
102886-380EA 561.11 USD
102886-380
Anti-NUDT5 Mouse Monoclonal Antibody [clone: 739044]
Antibodies
Type: Primary
Antigen: NUDT5
Clonality: Monoclonal
Clone: 739044
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR