Anti-EPHA4 Rat Monoclonal Antibody [clone: 107925]

Supplier: R & D Systems
MAB641
220021-385EA 763.57 USD
220021-385
Anti-EPHA4 Rat Monoclonal Antibody [clone: 107925]
Antibodies
Type: Primary
Antigen: EPHA4
Clonality: Monoclonal
Clone: 107925
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR