Anti-FUT7 Mouse Monoclonal Antibody [clone: 795116]

Supplier: R & D Systems
MAB64091
102886-378EA 594.37 USD
102886-378
Anti-FUT7 Mouse Monoclonal Antibody [clone: 795116]
Antibodies
Type: Primary
Antigen Symbol: FUT7
Antigen Name: fucosyltransferase 7 (alpha (1,3) fucosyltransferase)
Antigen Synonyms: FucT-VII
Clonality: Monoclonal
Clone: 795116
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR