Anti-AKT1S1 Mouse Monoclonal Antibody [clone: 66928]

Supplier: R & D Systems
MAB6408
102886-376EA 594.37 USD
102886-376
Anti-AKT1S1 Mouse Monoclonal Antibody [clone: 66928]
Antibodies
Simple Western

Type: Primary
Antigen: AKT1S1
Clonality: Monoclonal
Clone: 66928
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR