Anti-BAD Mouse Monoclonal Antibody [clone: 612312]

Supplier: R & D Systems
MAB6405
102886-372EA 594.37 USD
102886-372
Anti-BAD Mouse Monoclonal Antibody [clone: 612312]
Antibodies
Type: Primary
Antigen: BAD
Clonality: Monoclonal
Clone: 612312
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR