Anti-EPHA3 Rat Monoclonal Antibody [clone: 99437]

Supplier: R & D Systems
MAB640
220021-384EA 763.57 USD
220021-384
Anti-EPHA3 Rat Monoclonal Antibody [clone: 99437]
Antibodies
Type: Primary
Antigen: EPHA3
Clonality: Monoclonal
Clone: 99437
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR