Anti-DOK7 Mouse Monoclonal Antibody [clone: 651601]

Supplier: R & D Systems
MAB6398
102886-368EA 561.11 USD
102886-368
Anti-DOK7 Mouse Monoclonal Antibody [clone: 651601]
Antibodies
Type: Primary
Antigen: DOK7
Clonality: Monoclonal
Clone: 651601
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR