Anti-GKN1 Mouse Monoclonal Antibody [clone: 665701]

Supplier: R & D Systems
MAB6395
102886-366EA 566.89 USD
102886-366
Anti-GKN1 Mouse Monoclonal Antibody [clone: 665701]
Antibodies
Type: Primary
Antigen Symbol: GKN1
Antigen Name: gastrokine 1
Antigen Synonyms: AMP18, BRICD1, CA11, FOV, foveolin
Clonality: Monoclonal
Clone: 665701
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR