Anti-PIK3R5 Mouse Monoclonal Antibody [clone: 601214]

Supplier: R & D Systems
MAB6388
102886-362EA 577.02 USD
102886-362
Anti-PIK3R5 Mouse Monoclonal Antibody [clone: 601214]
Antibodies
Type: Primary
Antigen: PIK3R5
Clonality: Monoclonal
Clone: 601214
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR