Anti-SCTR Mouse Monoclonal Antibody [clone: 783526]

Supplier: R & D Systems
MAB6387
102886-360EA 577.02 USD
102886-360
Anti-SCTR Mouse Monoclonal Antibody [clone: 783526]
Antibodies
Type: Primary
Antigen: SCTR
Clonality: Monoclonal
Clone: 783526
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR