Anti-EPHA1 Mouse Monoclonal Antibody [clone: 86810]

Supplier: R & D Systems
MAB638
220021-382EA 763.57 USD
220021-382
Anti-EPHA1 Mouse Monoclonal Antibody [clone: 86810]
Antibodies
Type: Primary
Antigen: EPHA1
Clonality: Monoclonal
Clone: 86810
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR