Anti-CHRM2 Mouse Monoclonal Antibody [clone: 585713]

Supplier: R & D Systems
MAB6377
102885-364EA 584.25 USD
102885-364
Anti-CHRM2 Mouse Monoclonal Antibody [clone: 585713]
Antibodies
Type: Primary
Antigen: CHRM2
Clonality: Monoclonal
Clone: 585713
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR