Anti-SALL4 Mouse Monoclonal Antibody [clone: 651839]

Supplier: R & D Systems
MAB6374
102885-360EA 594.37 USD
102885-360
Anti-SALL4 Mouse Monoclonal Antibody [clone: 651839]
Antibodies
Type: Primary
Antigen: SALL4
Clonality: Monoclonal
Clone: 651839
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR