Anti-DDX5 Mouse Monoclonal Antibody [clone: 632813]

Supplier: R & D Systems
MAB6370
102885-358EA 577.02 USD
102885-358
Anti-DDX5 Mouse Monoclonal Antibody [clone: 632813]
Antibodies
Type: Primary
Antigen: DDX5
Clonality: Monoclonal
Clone: 632813
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR