Anti-CD302 Mouse Monoclonal Antibody [clone: 771910]

Supplier: R & D Systems
MAB6367
102885-356EA 594.37 USD
102885-356
Anti-CD302 Mouse Monoclonal Antibody [clone: 771910]
Antibodies
Type: Primary
Antigen: CD302
Clonality: Monoclonal
Clone: 771910
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR