Anti-LILRA2 Mouse Monoclonal Antibody [clone: 600007]

Supplier: R & D Systems
MAB6364
102885-354EA 594.37 USD
102885-354
Anti-LILRA2 Mouse Monoclonal Antibody [clone: 600007]
Antibodies
Type: Primary
Antigen: LILRA2
Clonality: Monoclonal
Clone: 600007
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR