Anti-LRP1 Mouse Monoclonal Antibody [clone: 545503]

Supplier: R & D Systems
MAB6360
102885-352EA 594.37 USD
102885-352
Anti-LRP1 Mouse Monoclonal Antibody [clone: 545503]
Antibodies
Type: Primary
Antigen: LRP1
Clonality: Monoclonal
Clone: 545503
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR