Anti-ZNF281 Mouse Monoclonal Antibody [clone: 801706]

Supplier: R & D Systems
MAB6350
102885-350EA 561.11 USD
102885-350
Anti-ZNF281 Mouse Monoclonal Antibody [clone: 801706]
Antibodies
Simple Western

Type: Primary
Antigen: ZNF281
Clonality: Monoclonal
Clone: 801706
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR