Anti-FGF4 Mouse Monoclonal Antibody [clone: 26019]

Supplier: R & D Systems
MAB635
220021-379EA 772.25 USD
220021-379
Anti-FGF4 Mouse Monoclonal Antibody [clone: 26019]
Antibodies
Type: Primary
Antigen Symbol: FGF4
Antigen Name: fibroblast growth factor 4
Antigen Synonyms: HBGF-4, HST, HST-1, HSTF1, K-FGF, KFGF
Clonality: Monoclonal
Clone: 26019
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR