Anti-TOP2B Mouse Monoclonal Antibody [clone: 681417]

Supplier: R & D Systems
MAB6348
102885-348EA 594.37 USD
102885-348
Anti-TOP2B Mouse Monoclonal Antibody [clone: 681417]
Antibodies
Type: Primary
Antigen: TOP2B
Clonality: Monoclonal
Clone: 681417
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR