Anti-IRS2 Mouse Monoclonal Antibody [clone: 676415]

Supplier: R & D Systems
MAB6347
102885-346EA 577.02 USD
102885-346
Anti-IRS2 Mouse Monoclonal Antibody [clone: 676415]
Antibodies
Type: Primary
Antigen: IRS2
Clonality: Monoclonal
Clone: 676415
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR