Anti-COL13A1 Mouse Monoclonal Antibody [clone: 584718]

Supplier: R & D Systems
MAB6346
102885-344EA 577.02 USD
102885-344
Anti-COL13A1 Mouse Monoclonal Antibody [clone: 584718]
Antibodies
Type: Primary
Antigen: COL13A1
Clonality: Monoclonal
Clone: 584718
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR