Anti-POR Mouse Monoclonal Antibody [clone: 739613]

Supplier: R & D Systems
MAB6340
102885-342EA 577.02 USD
102885-342
Anti-POR Mouse Monoclonal Antibody [clone: 739613]
Antibodies
Type: Primary
Antigen: POR
Clonality: Monoclonal
Clone: 739613
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR