Anti-CRTC2 Mouse Monoclonal Antibody [clone: 628430]

Supplier: R & D Systems
MAB6338
102885-340EA 577.02 USD
102885-340
Anti-CRTC2 Mouse Monoclonal Antibody [clone: 628430]
Antibodies
Type: Primary
Antigen: CRTC2
Clonality: Monoclonal
Clone: 628430
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR