Anti-TBX18 Mouse Monoclonal Antibody [clone: 635305]

Supplier: R & D Systems
MAB63371
102885-338EA 600.15 USD
102885-338
Anti-TBX18 Mouse Monoclonal Antibody [clone: 635305]
Antibodies
Type: Primary
Antigen: TBX18
Clonality: Monoclonal
Clone: 635305
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR