Anti-TBX18 Mouse Monoclonal Antibody [clone: 635303]

Supplier: R & D Systems
MAB6337
102885-336EA 594.37 USD
102885-336
Anti-TBX18 Mouse Monoclonal Antibody [clone: 635303]
Antibodies
Type: Primary
Antigen: TBX18
Clonality: Monoclonal
Clone: 635303
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR