Anti-HS2ST1 Mouse Monoclonal Antibody [clone: 738306]

Supplier: R & D Systems
MAB6335
102885-334EA 577.02 USD
102885-334
Anti-HS2ST1 Mouse Monoclonal Antibody [clone: 738306]
Antibodies
Type: Primary
Antigen: HS2ST1
Clonality: Monoclonal
Clone: 738306
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR