Anti-LAT Mouse Monoclonal Antibody [clone: 661002]

Supplier: R & D Systems
MAB63341
102885-332EA 594.37 USD
102885-332
Anti-LAT Mouse Monoclonal Antibody [clone: 661002]
Antibodies
Type: Primary
Antigen: LAT
Clonality: Monoclonal
Clone: 661002
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR