Anti-LAT Mouse Monoclonal Antibody [clone: 627945]

Supplier: R & D Systems
MAB6334
102885-330EA 577.02 USD
102885-330
Anti-LAT Mouse Monoclonal Antibody [clone: 627945]
Antibodies
Type: Primary
Antigen: LAT
Clonality: Monoclonal
Clone: 627945
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR