Anti-TNFRSF10D Mouse Monoclonal Antibody [clone: 104918]

Supplier: R & D Systems
MAB633
220021-377EA 779.48 USD
220021-377
Anti-TNFRSF10D Mouse Monoclonal Antibody [clone: 104918]
Antibodies
Type: Primary
Antigen: TNFRSF10D
Clonality: Monoclonal
Clone: 104918
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR