Anti-FOXC1 Mouse Monoclonal Antibody [clone: 625905]

Supplier: R & D Systems
MAB6329
102886-358EA 566.89 USD
102886-358
Anti-FOXC1 Mouse Monoclonal Antibody [clone: 625905]
Antibodies
Type: Primary
Antigen Symbol: FOXC1
Antigen Name: forkhead box C1
Antigen Synonyms: ARA, FKHL7, FREAC-3, FREAC3, IGDA, IHG1, IRID1, RIEG3
Clonality: Monoclonal
Clone: 625905
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR