Anti-BMP6 Rat Monoclonal Antibody [clone: 719040]

Supplier: R & D Systems
MAB6325
102886-356EA 594.37 USD
102886-356
Anti-BMP6 Rat Monoclonal Antibody [clone: 719040]
Antibodies
Type: Primary
Antigen: BMP6
Clonality: Monoclonal
Clone: 719040
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR