Anti-VGF Mouse Monoclonal Antibody [clone: 619707]

Supplier: R & D Systems
MAB6317
102886-354EA 594.37 USD
102886-354
Anti-VGF Mouse Monoclonal Antibody [clone: 619707]
Antibodies
Type: Primary
Antigen: VGF
Clonality: Monoclonal
Clone: 619707
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR