Anti-PIWIL4 Mouse Monoclonal Antibody [clone: 626001]

Supplier: R & D Systems
MAB6316
102886-352EA 577.02 USD
102886-352
Anti-PIWIL4 Mouse Monoclonal Antibody [clone: 626001]
Antibodies
Type: Primary
Antigen: PIWIL4
Clonality: Monoclonal
Clone: 626001
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR