Anti-DNMT3A Mouse Monoclonal Antibody [clone: 681615]

Supplier: R & D Systems
MAB63151
102886-350EA 616.06 USD
102886-350
Anti-DNMT3A Mouse Monoclonal Antibody [clone: 681615]
Antibodies
Type: Primary
Antigen: DNMT3A
Clonality: Monoclonal
Clone: 681615
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR