Anti-DNMT3A Mouse Monoclonal Antibody [clone: 681630]

Supplier: R & D Systems
MAB6315
102886-348EA 577.02 USD
102886-348
Anti-DNMT3A Mouse Monoclonal Antibody [clone: 681630]
Antibodies
Type: Primary
Antigen: DNMT3A
Clonality: Monoclonal
Clone: 681630
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR