Anti-TNFRSF10B Mouse Monoclonal Antibody [clone: 603307]

Supplier: R & D Systems
MAB6313
102886-346EA 577.02 USD
102886-346
Anti-TNFRSF10B Mouse Monoclonal Antibody [clone: 603307]
Antibodies
Type: Primary
Antigen: TNFRSF10B
Clonality: Monoclonal
Clone: 603307
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR